Assay Details
Gene Symbol: Pcsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017091.2 | NP_058787.1 | 1 - 2 | 398 | 94 | |
GenBank mRNA | M76705.1 | - | 1 - 2 | 400 | 94 | |
M83745.1 | - | 1 - 2 | 342 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
