Assay Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035233.1 | NP_001030310.1 | 272 | 60 | ||
| NM_017215.2 | NP_058911.2 | 272 | 60 | |||
| GenBank mRNA | AF465909.1 | - | 165 | 60 | ||
| AY044832.1 | - | 161 | 60 | |||
| CB760876.1 | - | 273 | 60 | |||
| JN132400.1 | - | 566 | 60 | |||
| U15098.1 | - | 221 | 60 | |||
| X67857.1 | - | 259 | 60 | |||
More Information
Set Membership: |
|

Slc1a2
Genomic Map