Assay Details
| Gene Symbol: Cyp2c11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019184.2 | NP_062057.2 | 3 - 4 | 505 | 65 | |
| XM_008760364.1 | XP_008758586.1 | 2 - 3 | 333 | 65 | ||
| GenBank mRNA | BC088146.1 | - | 3 - 4 | 495 | 7369647 | 65 |
| J02657.1 | - | 3 - 4 | 505 | 65 | ||
| U33173.1 | - | 3 - 4 | 505 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2c11
Genomic Map