Assay Details
| Gene Symbol: Sox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019193.2 | NP_062066.1 | 3 - 4 | 1015 | 114 | |
| GenBank mRNA | AJ001029.1 | - | 3 - 4 | 1016 | 114 | |
| BC062067.1 | - | 2 - 3 | 727 | 5599259 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sox10
Genomic Map