Assay Details
| Gene Symbol: Pak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019210.1 | NP_062083.1 | 6 - 7 | 652 | 72 | |
| XM_006257333.2 | XP_006257395.1 | 8 - 9 | 1925 | 72 | ||
| XM_006257336.2 | XP_006257398.1 | 7 - 8 | 1814 | 72 | ||
| XM_006257338.2 | XP_006257400.1 | 7 - 8 | 2217 | 72 | ||
| XM_006257339.2 | XP_006257401.1 | 7 - 8 | 1879 | 72 | ||
| XM_008773404.1 | XP_008771626.1 | 9 - 10 | 1990 | 72 | ||
| GenBank mRNA | U33314.1 | - | 6 - 7 | 652 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pak3
Genomic Map