Assay Details
| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020086.1 | NP_064471.1 | 3 - 4 | 1193 | 76 | |
| GenBank mRNA | AF154831.1 | - | 3 - 4 | 1193 | 76 | |
| BC070900.1 | - | 3 - 4 | 1193 | 7100403 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plvap
Genomic Map