Assay Details
Gene Symbol: LOC100909712 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749295.3 | XP_003749343.1 | 1 - 2 | 706 | 97 |
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053662.3 | NP_446114.1 | 397 | 97 | ||
GenBank mRNA | AF030091.3 | - | 397 | 97 | ||
BC085686.1 | - | 378 | 97 | |||
BC128699.1 | - | 344 | 97 | |||
BC161817.1 | - | 343 | 9022416 | 97 | ||
CB608234.1 | - | 404 | 97 | |||
FQ232436.1 | - | 361 | 97 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
