Assay Details
| Gene Symbol: Sdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053893.3 | NP_446345.3 | 2 - 3 | 297 | 101 | |
| GenBank mRNA | U73184.1 | - | 2 - 3 | 365 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdc3
Genomic Map