Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177930.2 | NP_808791.1 | 18 - 19 | 2469 | 91 | |
| GenBank mRNA | AY220476.1 | - | 18 - 19 | 2469 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P148720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87875 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P148720 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P148720 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P148720 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P222292 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P148720 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51336 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94859 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93168 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37922 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9876 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44101 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10807991 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807984 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807989 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807978 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807980 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807990 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807982 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807986 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807988 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10807975 | - | - |
| Rat Genome 230 2.0 Array | 1389041_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1396926_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389041_at | - | - |
| Rat Genome 230 2.0 Array | 1396926_at | - | - |
| Affymetrix HT Rat Focus Array | 1389041_at | - | - |
| Affymetrix HT Rat Focus Array | 1389041_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1396926_at | - | - |
| Affymetrix HT Rat Focus Array | 1396926_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900011444.rn.1 | TC1900001411.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389041_at | - | - |
| Rat Expression Set 230 | 1389041_PM_at | - | - |
| Rat Expression Set 230 | 1396926_at | - | - |
| Rat Expression Set 230 | 1396926_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10807975 | - | - |
| Rat Genome U34 Set | rc_AI008813_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21315805 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359871 | - | - |
| RatRef-12 Expression BeadChip | ILMN_53347 | - | - |

Vac14
Genomic Map