Assay Details
Gene Symbol: Sel1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177933.2 | NP_808794.2 | 6 - 7 | 869 | 60 | |
GenBank mRNA | AF304853.1 | - | 6 - 7 | 883 | 60 | |
AF304855.1 | - | 2 - 3 | 268 | 60 | ||
CB814385.1 | - | 3 - 4 | 304 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P125958 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P517576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108693 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P517576 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P125958 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P517576 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P125958 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P125958 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P517576 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P517576 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53966 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34857 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97934 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29010 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10891460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891476 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891477 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891455 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891463 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891467 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891459 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891464 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891468 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891457 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891472 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891466 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891471 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891454 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891473 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891465 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891470 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891462 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10891453 | - | - |
Rat Genome 230 2.0 Array | 1373313_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392955_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373313_at | - | - |
Rat Genome 230 2.0 Array | 1392955_at | - | - |
Rat Genome 230 2.0 Array | 1376073_at | - | - |
Rat Genome 230 2.0 Array | 1376073_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373313_at | - | - |
Affymetrix HT Rat Focus Array | 1376073_at | - | - |
Affymetrix HT Rat Focus Array | 1392955_at | - | - |
Affymetrix HT Rat Focus Array | 1376073_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373313_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392955_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600020904.rn.1 | TC0600002950.rn.1 | RTA TC |
Rat Expression Set 230 | 1376073_PM_at | - | - |
Rat Expression Set 230 | 1376073_at | - | - |
Rat Expression Set 230 | 1373313_PM_at | - | - |
Rat Expression Set 230 | 1373313_at | - | - |
Rat Expression Set 230 | 1392955_at | - | - |
Rat Expression Set 230 | 1392955_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891453 | - | - |
Rat Genome U34 Set | rc_AA859897_at | - | - |
Rat Genome U34 Set | rc_AI072921_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20721657 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350394 | - | - |
RatRef-12 Expression BeadChip | ILMN_63090 | - | - |
