Assay Details
| Gene Symbol: Egln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772679.1 | XP_008770901.1 | 5 - 6 | 1765 | 65 | |
| GenBank mRNA | AY228140.1 | - | 3 - 4 | 550 | 65 | |
| BC081694.1 | - | 3 - 4 | 794 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Egln1
Genomic Map