Assay Details
| Gene Symbol: Syngap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113409.2 | NP_001106880.2 | 11 - 12 | 2299 | 68 | |
| NM_181092.4 | NP_851606.3 | 13 - 14 | 2362 | 68 | ||
| GenBank mRNA | AF048976.1 | - | 13 - 14 | 2370 | 68 | |
| AF058789.2 | - | 13 - 14 | 2249 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syngap1
Genomic Map