Assay Details
Gene Symbol: Adam28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181693.1 | NP_859044.1 | 18 - 19 | 2025 | 72 | |
XM_008770820.1 | XP_008769042.1 | 18 - 19 | 2036 | 72 | ||
GenBank mRNA | AY283187.1 | - | 18 - 19 | 2025 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P468147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170372 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P468147 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P468147 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P468147 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P468147 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48946 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66369 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50673 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29408 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10784888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784880 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784876 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784877 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784891 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784889 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784873 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784887 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784886 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784883 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784874 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784890 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784879 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10784866 | - | - |
Affymetrix Rat Clariom S Assay | PSR1500013333.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013335.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013339.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013342.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013336.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013345.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013348.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013349.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013344.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013350.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013331.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013346.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013334.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013338.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013341.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013343.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013347.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013337.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500013340.rn.1 | TC1500001960.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10784866 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20872547 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360745 | - | - |
RatRef-12 Expression BeadChip | ILMN_49800 | - | - |
