Assay Details
Gene Symbol: Vkorc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203335.2 | NP_976080.1 | 1 - 2 | 200 | 55 | |
GenBank mRNA | AY423047.1 | - | 1 - 2 | 183 | 55 | |
BC166413.1 | - | 196 | 9100951 | 55 | ||
CB314647.1 | - | 1 - 2 | 216 | 55 | ||
FQ210919.1 | - | 1 - 2 | 242 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
