Assay Details
| Gene Symbol: Kcnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013186.1 | NP_037318.1 | 1 - 2 | 575 | 75 | |
| GenBank mRNA | X16476.1 | - | 1 - 2 | 575 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
regulation of ion transmembrane transport
protein oligomerization
protein homooligomerization
potassium ion transmembrane transport
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
outward rectifier potassium channel activity
protein N-terminus binding
potassium ion transport
regulation of ion transmembrane transport
protein oligomerization
protein homooligomerization
potassium ion transmembrane transport
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
outward rectifier potassium channel activity
protein N-terminus binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P675031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153954 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P675031 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P675031 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P675031 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P675031 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701914521 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49658 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32684 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51987 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1153 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35575 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59633 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701914521 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10851946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10851944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10851945 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10851943 | - | - |
| Rat Genome 230 2.0 Array | 1368242_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368242_at | - | - |
| Affymetrix HT Rat Focus Array | 1368242_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368242_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300037015.rn.1 | TC0300004817.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300037013.rn.1 | TC0300004817.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300037014.rn.1 | TC0300004817.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368242_PM_at | - | - |
| Rat Expression Set 230 | 1368242_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10851943 | - | - |
| Rat Genome U34 Set | X16476cds_at | - | - |
| Rat Genome U34 Set | X16476cds_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20889867 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359651 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52681 | - | - |

Kcnb1
Genomic Map