Assay Details
Gene Symbol: Kcnj6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013192.2 | NP_037324.1 | 2 - 3 | 1164 | 70 | |
XM_008768567.1 | XP_008766789.1 | 1 - 2 | 950 | 70 | ||
GenBank mRNA | AB073753.1 | - | 2 - 3 | 1164 | 70 | |
AB073754.1 | - | 2 - 3 | 1164 | 70 | ||
AB073755.1 | - | 2 - 3 | 1164 | 70 | ||
EF156275.1 | - | 1 - 2 | 303 | 70 | ||
U21087.1 | - | 2 - 3 | 1164 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion import
regulation of ion transmembrane transport
thyroid hormone metabolic process
response to electrical stimulus
cellular response to morphine
potassium ion transmembrane transport
G-protein alpha-subunit binding
protein binding
G-protein activated inward rectifier potassium channel activity
potassium ion import
regulation of ion transmembrane transport
thyroid hormone metabolic process
response to electrical stimulus
cellular response to morphine
potassium ion transmembrane transport
G-protein alpha-subunit binding
protein binding
G-protein activated inward rectifier potassium channel activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P182270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4225 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P182270 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P182270 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P182270 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P208113 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P182270 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57519 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62984 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97949 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15876 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1272 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15097 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8130 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10753275 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753277 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753270 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753278 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753273 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753276 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753271 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753272 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753274 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10753269 | - | - |
Rat Genome 230 2.0 Array | 1370704_at | - | - |
Rat Genome 230 2.0 Array | 1370598_a_at | - | - |
Rat Genome 230 2.0 Array | 1369035_a_at | - | - |
Rat Genome 230 2.0 Array | 1370704_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370598_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1369034_at | - | - |
Rat Genome 230 2.0 Array | 1369034_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369035_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370704_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369034_at | - | - |
Affymetrix HT Rat Focus Array | 1370704_at | - | - |
Affymetrix HT Rat Focus Array | 1369035_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369035_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369034_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370598_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370598_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100008149.rn.1 | TC1100001366.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100008148.rn.1 | TC1100001366.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100008154.rn.1 | TC1100001366.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100008150.rn.1 | TC1100001366.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100008152.rn.1 | TC1100001366.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100008153.rn.1 | TC1100001366.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100008155.rn.1 | TC1100001366.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100008147.rn.1 | TC1100001366.rn.1 | RTA TC |
Rat Expression Set 230 | 1370704_PM_at | - | - |
Rat Expression Set 230 | 1369034_at | - | - |
Rat Expression Set 230 | 1370598_a_at | - | - |
Rat Expression Set 230 | 1370598_PM_a_at | - | - |
Rat Expression Set 230 | 1370704_at | - | - |
Rat Expression Set 230 | 1369035_PM_a_at | - | - |
Rat Expression Set 230 | 1369035_a_at | - | - |
Rat Expression Set 230 | 1369034_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753269 | - | - |
Rat Genome U34 Set | U21087_at | - | - |
Rat Genome U34 Set | U21087_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21923345 | - | - |
