Assay Details
| Gene Symbol: Csrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017148.2 | NP_058844.1 | 2 - 3 | 153 | 61 | |
| GenBank mRNA | BC062407.1 | - | 2 - 3 | 153 | 5623423 | 61 |
| U09567.1 | - | 2 - 3 | 148 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csrp1
Genomic Map