Assay Details
| Gene Symbol: Ces1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031565.2 | NP_113753.2 | 3 | 604 | 72 | |
| GenBank mRNA | EX493673.1 | - | 3 - 4 | 396 | 72 | |
| X81395.1 | - | 3 | 469 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ces1e
Genomic Map