Assay Details
Gene Symbol: Tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145778.2 | NP_665721.1 | 5 - 6 | 583 | 67 | |
GenBank mRNA | AB015946.1 | - | 5 - 6 | 488 | 67 | |
BC070957.1 | - | 5 - 6 | 583 | 7105643 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
