Assay Details
Gene Symbol: Ston1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182954.2 | NP_891999.2 | 2 - 3 | 2247 | 82 | |
XM_006239743.2 | XP_006239805.1 | 2 - 3 | 2018 | 82 | ||
XM_006239744.2 | XP_006239806.1 | 2 - 3 | 3114 | 82 | ||
XM_008764479.1 | XP_008762701.1 | 3 - 4 | 2335 | 82 | ||
XM_008764480.1 | XP_008762702.1 | 2 - 3 | 2803 | 82 | ||
XM_008764481.1 | XP_008762703.1 | 2 - 3 | 1998 | 82 | ||
GenBank mRNA | AY300793.1 | - | 1 - 2 | 1959 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
