Assay Details
| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183331.1 | NP_899160.1 | 25 - 26 | 6699 | 63 | |
| XM_008771944.1 | XP_008770166.1 | 9 - 10 | 1167 | 63 | ||
| XM_008771945.1 | XP_008770167.1 | 7 - 8 | 889 | 63 | ||
| GenBank mRNA | AY362193.1 | - | 25 - 26 | 6699 | 63 | |
| GU320196.1 | - | 25 - 26 | 6675 | 63 | ||
| GU320197.1 | - | 25 - 26 | 6675 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P057165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14959 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P226601 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P226601 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P226601 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P226601 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89297 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19351 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19100 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10800040 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800053 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800057 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800062 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800046 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800059 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800063 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800041 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800061 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800054 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800055 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800045 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800060 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800039 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800052 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800042 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800048 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800043 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800050 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800065 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800047 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800049 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800064 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10800038 | - | - |
| Rat Genome 230 2.0 Array | 1380190_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395015_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380367_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382194_at | - | - |
| Rat Genome 230 2.0 Array | 1396181_at | - | - |
| Rat Genome 230 2.0 Array | 1395916_at | - | - |
| Rat Genome 230 2.0 Array | 1396181_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382194_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395015_at | - | - |
| Rat Genome 230 2.0 Array | 1395916_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380367_at | - | - |
| Rat Genome 230 2.0 Array | 1380190_at | - | - |
| Affymetrix HT Rat Focus Array | 1395916_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382194_at | - | - |
| Affymetrix HT Rat Focus Array | 1380367_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380190_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380190_at | - | - |
| Affymetrix HT Rat Focus Array | 1396181_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395916_at | - | - |
| Affymetrix HT Rat Focus Array | 1395015_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395015_at | - | - |
| Affymetrix HT Rat Focus Array | 1396181_at | - | - |
| Affymetrix HT Rat Focus Array | 1382194_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380367_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800007059.rn.1 | TC1800001000.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800007058.rn.1 | TC1800001000.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800007057.rn.1 | TC1800001000.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382194_PM_at | - | - |
| Rat Expression Set 230 | 1380190_PM_at | - | - |
| Rat Expression Set 230 | 1396181_PM_at | - | - |
| Rat Expression Set 230 | 1395916_at | - | - |
| Rat Expression Set 230 | 1380367_PM_at | - | - |
| Rat Expression Set 230 | 1380367_at | - | - |
| Rat Expression Set 230 | 1395916_PM_at | - | - |
| Rat Expression Set 230 | 1396181_at | - | - |
| Rat Expression Set 230 | 1380190_at | - | - |
| Rat Expression Set 230 | 1395015_at | - | - |
| Rat Expression Set 230 | 1382194_at | - | - |
| Rat Expression Set 230 | 1395015_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800038 | - | - |
| Rat Genome U34 Set | rc_AI176907_at | - | - |
| Rat Genome U34 Set | rc_AI234173_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21397473 | - | - |

F8
Genomic Map