Assay Details
| Gene Symbol: Eif5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100697.2 | NP_001094167.1 | 2 - 3 | 279 | 86 | |
| XM_006232231.2 | XP_006232293.1 | 2 - 3 | 1310 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif5a2
Genomic Map