Assay Details
| Gene Symbol: Akr1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012498.1 | NP_036630.1 | 5 - 6 | 585 | 99 | |
| GenBank mRNA | BC062034.1 | - | 5 - 6 | 570 | 5623700 | 99 |
| X05884.1 | - | 5 - 6 | 582 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Akr1b1
Genomic Map