Assay Details
| Gene Symbol: Ldha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017025.1 | NP_058721.1 | 3 - 4 | 350 | 85 | |
| XM_006229232.2 | XP_006229294.1 | 3 - 4 | 518 | 85 | ||
| GenBank mRNA | BC060587.1 | - | 3 - 4 | 302 | 6919582 | 85 |
| BC084698.1 | - | 2 - 3 | 252 | 7122339 | 85 | |
| BC168737.1 | - | 3 - 4 | 349 | 6917937 | 85 | |
| FQ214854.1 | - | 3 - 4 | 350 | 85 | ||
| X01964.1 | - | 3 - 4 | 350 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ldha
Genomic Map