Assay Details
| Gene Symbol: Rps3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017153.1 | NP_058849.1 | 4 - 5 | 587 | 82 | |
| GenBank mRNA | BC058483.1 | - | 4 - 5 | 592 | 6887884 | 82 |
| FQ211699.1 | - | 4 - 5 | 588 | 82 | ||
| FQ211708.1 | - | 4 - 5 | 587 | 82 | ||
| FQ216850.1 | - | 4 - 5 | 587 | 82 | ||
| FQ217809.1 | - | 4 - 5 | 587 | 82 | ||
| FQ219020.1 | - | 4 - 5 | 587 | 82 | ||
| FQ219522.1 | - | 4 - 5 | 588 | 82 | ||
| FQ219543.1 | - | 4 - 5 | 82 | |||
| FQ219867.1 | - | 4 - 5 | 588 | 82 | ||
| FQ220002.1 | - | 4 - 5 | 587 | 82 | ||
| FQ220027.1 | - | 4 - 5 | 587 | 82 | ||
| FQ220045.1 | - | 4 - 5 | 587 | 82 | ||
| FQ220135.1 | - | 4 - 5 | 588 | 82 | ||
| FQ220260.1 | - | 4 - 5 | 587 | 82 | ||
| FQ220261.1 | - | 4 - 5 | 588 | 82 | ||
| FQ220317.1 | - | 4 - 5 | 587 | 82 | ||
| FQ220504.1 | - | 4 - 5 | 587 | 82 | ||
| FQ220529.1 | - | 4 - 5 | 587 | 82 | ||
| FQ220577.1 | - | 4 - 5 | 588 | 82 | ||
| FQ220731.1 | - | 4 - 5 | 587 | 82 | ||
| FQ221431.1 | - | 4 - 5 | 588 | 82 | ||
| FQ221460.1 | - | 4 - 5 | 587 | 82 | ||
| FQ221744.1 | - | 4 - 5 | 587 | 82 | ||
| FQ221931.1 | - | 4 - 5 | 587 | 82 | ||
| FQ222056.1 | - | 4 - 5 | 587 | 82 | ||
| FQ222736.1 | - | 4 - 5 | 588 | 82 | ||
| FQ222773.1 | - | 4 - 5 | 588 | 82 | ||
| FQ222846.1 | - | 4 - 5 | 617 | 82 | ||
| FQ222869.1 | - | 4 - 5 | 587 | 82 | ||
| FQ222876.1 | - | 4 - 5 | 587 | 82 | ||
| FQ223535.1 | - | 4 - 5 | 594 | 82 | ||
| FQ223540.1 | - | 4 - 5 | 587 | 82 | ||
| FQ225080.1 | - | 4 - 5 | 588 | 82 | ||
| FQ225145.1 | - | 4 - 5 | 587 | 82 | ||
| FQ225713.1 | - | 4 - 5 | 586 | 82 | ||
| FQ227696.1 | - | 4 - 5 | 587 | 82 | ||
| FQ227805.1 | - | 4 - 5 | 588 | 82 | ||
| FQ227878.1 | - | 4 - 5 | 588 | 82 | ||
| FQ228312.1 | - | 4 - 5 | 588 | 82 | ||
| FQ228317.1 | - | 4 - 5 | 587 | 82 | ||
| FQ228376.1 | - | 4 - 5 | 588 | 82 | ||
| FQ228558.1 | - | 4 - 5 | 587 | 82 | ||
| FQ229234.1 | - | 4 - 5 | 587 | 82 | ||
| FQ229381.1 | - | 4 - 5 | 588 | 82 | ||
| FQ229520.1 | - | 4 - 5 | 587 | 82 | ||
| FQ229870.1 | - | 4 - 5 | 587 | 82 | ||
| FQ229960.1 | - | 4 - 5 | 587 | 82 | ||
| FQ231162.1 | - | 4 - 5 | 588 | 82 | ||
| FQ231673.1 | - | 4 - 5 | 587 | 82 | ||
| FQ232329.1 | - | 4 - 5 | 589 | 82 | ||
| FQ232836.1 | - | 4 - 5 | 587 | 82 | ||
| FQ232928.1 | - | 4 - 5 | 588 | 82 | ||
| FQ233768.1 | - | 4 - 5 | 591 | 82 | ||
| FQ234311.1 | - | 4 - 5 | 587 | 82 | ||
| FQ234537.1 | - | 4 - 5 | 587 | 82 | ||
| M84716.1 | - | 4 - 5 | 581 | 82 | ||
| X75161.1 | - | 4 - 5 | 587 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps3a
Genomic Map