Assay Details
| Gene Symbol: Rps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017160.1 | NP_058856.1 | 514 | 79 | ||
| GenBank mRNA | BC058149.1 | - | 544 | 6918741 | 79 | |
| FQ210697.1 | - | 146 | 79 | |||
| FQ211324.1 | - | 146 | 79 | |||
| FQ212249.1 | - | 146 | 79 | |||
| FQ217720.1 | - | 146 | 79 | |||
| FQ218111.1 | - | 146 | 79 | |||
| FQ221402.1 | - | 146 | 79 | |||
| FQ222806.1 | - | 146 | 79 | |||
| FQ229168.1 | - | 146 | 79 | |||
| FQ230362.1 | - | 146 | 79 | |||
| FQ231606.1 | - | 146 | 79 | |||
| M29358.1 | - | 514 | 79 | |||
| Gene Symbol: LOC100911372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749971.3 | XP_003750019.1 | 4 - 5 | 599 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps6
Genomic Map