Assay Details
| Gene Symbol: Hnrnpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017248.1 | NP_058944.1 | 9 - 10 | 998 | 77 | |
| XM_006242384.1 | XP_006242446.1 | 10 - 11 | 1495 | 82 | ||
| GenBank mRNA | BC062235.1 | - | 9 - 10 | 1014 | 6920834 | 82 |
| M12156.1 | - | 9 - 10 | 998 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P067163 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P944757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31287 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067163 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P944757 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P944757 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067163 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P944757 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P067163 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P944758 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P944757 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P485694 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P283372 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700244874 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700244874 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10899626 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932741 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899620 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899629 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899625 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798581 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932743 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899623 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899631 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899632 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899628 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899622 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899630 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932742 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899621 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798582 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899624 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10932740 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10798580 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10899619 | - | - |
| Rat Genome 230 2.0 Array | 1375612_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387872_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387872_at | - | - |
| Rat Genome 230 2.0 Array | 1375612_at | - | - |
| Rat Genome 230 2.0 Array | 1372693_at | - | - |
| Rat Genome 230 2.0 Array | 1372693_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387872_at | - | - |
| Affymetrix HT Rat Focus Array | 1372693_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372693_at | - | - |
| Affymetrix HT Rat Focus Array | 1387872_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700015122.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015124.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015137.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015126.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015130.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015128.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015138.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015143.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015141.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015125.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015127.rn.1 | TC0700001931.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015142.rn.1 | TC0700001931.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387872_PM_at | - | - |
| Rat Expression Set 230 | 1375612_PM_at | - | - |
| Rat Expression Set 230 | 1372693_at | - | - |
| Rat Expression Set 230 | 1372693_PM_at | - | - |
| Rat Expression Set 230 | 1375612_at | - | - |
| Rat Expression Set 230 | 1387872_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932740 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798580 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899619 | - | - |
| Rat Genome U34 Set | rc_AA965147_at | - | - |
| Rat Genome U34 Set | rc_AA799893_at | - | - |
| Rat Genome U34 Set | M12156_at | - | - |
| Rat Genome U34 Set | rc_AA799893_g_at | - | - |
| Rat Genome U34 Set | rc_AI237751_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22249880 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373050 | - | - |
| RatRef-12 Expression BeadChip | ILMN_47863 | - | - |

Hnrnpa1
Genomic Map