Assay Details
| Gene Symbol: Calm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC058485.1 | - | 74 | 6921326 | 71 | |
| FQ211494.1 | - | 74 | 71 | |||
| FQ211970.1 | - | 70 | 71 | |||
| FQ212357.1 | - | 71 | 71 | |||
| FQ212579.1 | - | 70 | 71 | |||
| FQ212640.1 | - | 70 | 71 | |||
| FQ212691.1 | - | 70 | 71 | |||
| FQ212789.1 | - | 74 | 71 | |||
| FQ212869.1 | - | 74 | 71 | |||
| FQ213028.1 | - | 70 | 71 | |||
| FQ213029.1 | - | 68 | 71 | |||
| FQ213091.1 | - | 70 | 71 | |||
| FQ213115.1 | - | 76 | 71 | |||
| FQ213200.1 | - | 73 | 71 | |||
| FQ213216.1 | - | 70 | 71 | |||
| FQ213366.1 | - | 69 | 71 | |||
| FQ213496.1 | - | 73 | 71 | |||
| FQ213520.1 | - | 74 | 71 | |||
| FQ213526.1 | - | 70 | 71 | |||
| FQ213581.1 | - | 70 | 71 | |||
| FQ213598.1 | - | 70 | 71 | |||
| FQ213637.1 | - | 70 | 71 | |||
| FQ213695.1 | - | 70 | 71 | |||
| FQ213703.1 | - | 70 | 71 | |||
| FQ213760.1 | - | 70 | 71 | |||
| FQ213762.1 | - | 75 | 71 | |||
| FQ213913.1 | - | 70 | 71 | |||
| FQ213941.1 | - | 74 | 71 | |||
| FQ213981.1 | - | 71 | 71 | |||
| FQ214059.1 | - | 74 | 71 | |||
| FQ214181.1 | - | 74 | 71 | |||
| FQ214262.1 | - | 74 | 71 | |||
| FQ214491.1 | - | 70 | 71 | |||
| FQ214500.1 | - | 70 | 71 | |||
| FQ214519.1 | - | 70 | 71 | |||
| FQ214540.1 | - | 74 | 71 | |||
| FQ215159.1 | - | 74 | 71 | |||
| FQ215317.1 | - | 70 | 71 | |||
| FQ215600.1 | - | 74 | 71 | |||
| FQ216397.1 | - | 70 | 71 | |||
| FQ216471.1 | - | 71 | 71 | |||
| FQ218091.1 | - | 70 | 71 | |||
| FQ220250.1 | - | 70 | 71 | |||
| FQ220325.1 | - | 70 | 71 | |||
| FQ220698.1 | - | 74 | 71 | |||
| FQ220779.1 | - | 70 | 71 | |||
| FQ222184.1 | - | 73 | 71 | |||
| FQ223357.1 | - | 74 | 71 | |||
| FQ226994.1 | - | 74 | 71 | |||
| FQ229228.1 | - | 70 | 71 | |||
| FQ229580.1 | - | 74 | 71 | |||
| FQ229619.1 | - | 70 | 71 | |||
| FQ229790.1 | - | 70 | 71 | |||
| FQ231022.1 | - | 74 | 71 | |||
| FQ233337.1 | - | 73 | 71 | |||
| FQ234377.1 | - | 74 | 71 | |||
| FQ235266.1 | - | 71 | 71 | |||
| M17069.1 | - | 72 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058485.1,M17069.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Calm2
Genomic Map