Assay Details
Gene Symbol: Timm23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019352.2 | NP_062225.1 | 3 - 4 | 361 | 61 | |
GenBank mRNA | AB006451.1 | - | 3 - 4 | 361 | 61 | |
BC083559.1 | - | 3 - 4 | 368 | 7135081 | 61 | |
FQ216304.1 | - | 3 - 4 | 365 | 61 | ||
FQ229692.1 | - | 3 - 4 | 365 | 61 | ||
FQ229803.1 | - | 3 - 4 | 367 | 61 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
