Assay Details
| Gene Symbol: Timm23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019352.2 | NP_062225.1 | 4 - 5 | 439 | 62 | |
| GenBank mRNA | AB006451.1 | - | 4 - 5 | 439 | 62 | |
| BC083559.1 | - | 4 - 5 | 446 | 7135081 | 62 | |
| FQ216304.1 | - | 4 - 5 | 443 | 62 | ||
| FQ229692.1 | - | 4 - 5 | 443 | 62 | ||
| FQ229803.1 | - | 4 - 5 | 445 | 62 | ||
| Gene Symbol: LOC100362432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729199.4 | XP_002729245.1 | 1 | 552 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Timm23
Genomic Map