Assay Details
| Gene Symbol: Rpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022514.1 | NP_071959.1 | 3 - 4 | 383 | 69 | |
| GenBank mRNA | BC058474.1 | - | 4 - 5 | 413 | 6919338 | 69 |
| BC091566.1 | - | 4 - 5 | 381 | 7388530 | 69 | |
| FQ210011.1 | - | 4 - 5 | 399 | 69 | ||
| FQ214023.1 | - | 4 - 5 | 403 | 69 | ||
| FQ217713.1 | - | 3 - 4 | 323 | 69 | ||
| FQ221292.1 | - | 4 - 5 | 408 | 69 | ||
| FQ221643.1 | - | 4 - 5 | 400 | 69 | ||
| FQ221684.1 | - | 4 - 5 | 401 | 69 | ||
| FQ221938.1 | - | 4 - 5 | 414 | 69 | ||
| FQ222018.1 | - | 4 - 5 | 403 | 69 | ||
| FQ222606.1 | - | 4 - 5 | 404 | 69 | ||
| FQ222991.1 | - | 4 - 5 | 405 | 69 | ||
| FQ223942.1 | - | 3 - 4 | 332 | 69 | ||
| FQ224664.1 | - | 4 - 5 | 401 | 69 | ||
| FQ228505.1 | - | 4 - 5 | 400 | 69 | ||
| FQ228603.1 | - | 4 - 5 | 401 | 69 | ||
| FQ228786.1 | - | 4 - 5 | 400 | 69 | ||
| X07424.1 | - | 3 - 4 | 383 | 69 | ||
More Information
Set Membership: |
|

Rpl27
Genomic Map