Assay Details
| Gene Symbol: Ube2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022690.2 | NP_073181.1 | 2 - 3 | 456 | 145 | |
| GenBank mRNA | AF099093.1 | - | 2 - 3 | 423 | 145 | |
| BC086980.1 | - | 2 - 3 | 456 | 7192141 | 145 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ube2g1
Genomic Map