Assay Details
| Gene Symbol: Apex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024148.1 | NP_077062.1 | 2 - 3 | 160 | 86 | |
| XM_006251913.1 | XP_006251975.1 | 2 - 3 | 201 | 86 | ||
| GenBank mRNA | AF309114.1 | - | 1 - 2 | 76 | 86 | |
| AF311054.1 | - | 1 - 2 | 38 | 86 | ||
| BC078816.1 | - | 2 - 3 | 168 | 7127205 | 86 | |
| D44495.1 | - | 2 - 3 | 160 | 86 | ||
| FQ220521.1 | - | 2 - 3 | 162 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Apex1
Genomic Map