Assay Details
| Gene Symbol: Rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031100.1 | NP_112362.1 | 3 - 4 | 144 | 77 | |
| GenBank mRNA | BC058467.1 | - | 3 - 4 | 133 | 6918973 | 77 |
| FQ210278.1 | - | 3 - 4 | 128 | 77 | ||
| FQ220555.1 | - | 3 - 4 | 127 | 77 | ||
| FQ221864.1 | - | 3 - 4 | 128 | 77 | ||
| FQ222021.1 | - | 3 - 4 | 128 | 77 | ||
| FQ229032.1 | - | 3 - 4 | 127 | 77 | ||
| FQ229696.1 | - | 1 - 2 | 255 | 77 | ||
| X87106.1 | - | 3 - 4 | 144 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl10
Genomic Map