Assay Details
| Gene Symbol: Rpl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031102.2 | NP_112364.1 | 2 - 3 | 110 | 84 | |
| XM_006229161.2 | XP_006229223.1 | 2 - 3 | 591 | 84 | ||
| GenBank mRNA | BC084727.1 | - | 1 - 2 | 102 | 7308070 | 84 |
| CB577548.1 | - | 2 - 3 | 110 | 84 | ||
| FQ211765.1 | - | 2 - 3 | 128 | 84 | ||
| FQ221110.1 | - | 2 - 3 | 128 | 84 | ||
| FQ222651.1 | - | 2 - 3 | 128 | 84 | ||
| FQ228615.1 | - | 2 - 3 | 131 | 84 | ||
| M20156.1 | - | 1 - 2 | 96 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl18
Genomic Map