Assay Details
| Gene Symbol: Pgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053291.3 | NP_445743.2 | 8 - 9 | 942 | 84 | |
| GenBank mRNA | BC063161.1 | - | 8 - 9 | 1023 | 6921844 | 84 |
| BC087651.1 | - | 8 - 9 | 942 | 7310274 | 84 | |
| FQ224830.1 | - | 3 - 4 | 435 | 84 | ||
| M31788.1 | - | 8 - 9 | 978 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pgk1
Genomic Map