Assay Details
Gene Symbol: Gnb2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130734.2 | NP_570090.2 | 1 - 2 | 245 | 67 | |
GenBank mRNA | BC063809.1 | - | 1 - 2 | 220 | 6919982 | 67 |
FQ212638.1 | - | 1 - 2 | 216 | 67 | ||
FQ213067.1 | - | 1 - 2 | 216 | 67 | ||
FQ213571.1 | - | 1 - 2 | 217 | 67 | ||
FQ215527.1 | - | 1 - 2 | 216 | 67 | ||
FQ216167.1 | - | 1 - 2 | 216 | 67 | ||
FQ218475.1 | - | 1 - 2 | 246 | 67 | ||
FQ218650.1 | - | 1 - 2 | 220 | 67 | ||
FQ219434.1 | - | 1 - 2 | 216 | 67 | ||
FQ219492.1 | - | 1 - 2 | 217 | 67 | ||
FQ219693.1 | - | 1 - 2 | 216 | 67 | ||
FQ219729.1 | - | 1 - 2 | 216 | 67 | ||
FQ219878.1 | - | 1 - 2 | 214 | 67 | ||
FQ219900.1 | - | 1 - 2 | 216 | 67 | ||
FQ220124.1 | - | 1 - 2 | 216 | 67 | ||
FQ220151.1 | - | 1 - 2 | 216 | 67 | ||
FQ220277.1 | - | 1 - 2 | 216 | 67 | ||
FQ220314.1 | - | 1 - 2 | 216 | 67 | ||
FQ220419.1 | - | 1 - 2 | 216 | 67 | ||
FQ220428.1 | - | 1 - 2 | 220 | 67 | ||
FQ220430.1 | - | 1 - 2 | 215 | 67 | ||
FQ220472.1 | - | 1 - 2 | 216 | 67 | ||
FQ220494.1 | - | 1 - 2 | 216 | 67 | ||
FQ220585.1 | - | 1 - 2 | 216 | 67 | ||
FQ220598.1 | - | 1 - 2 | 220 | 67 | ||
FQ220697.1 | - | 1 - 2 | 220 | 67 | ||
FQ220819.1 | - | 1 - 2 | 217 | 67 | ||
FQ220976.1 | - | 1 - 2 | 223 | 67 | ||
FQ221075.1 | - | 1 - 2 | 216 | 67 | ||
FQ221140.1 | - | 1 - 2 | 223 | 67 | ||
FQ221286.1 | - | 1 - 2 | 220 | 67 | ||
FQ222502.1 | - | 1 - 2 | 216 | 67 | ||
FQ223855.1 | - | 1 - 2 | 215 | 67 | ||
FQ224952.1 | - | 1 - 2 | 216 | 67 | ||
FQ225047.1 | - | 1 - 2 | 220 | 67 | ||
FQ225312.1 | - | 1 - 2 | 215 | 67 | ||
FQ225511.1 | - | 1 - 2 | 216 | 67 | ||
FQ225843.1 | - | 1 - 2 | 246 | 67 | ||
FQ226802.1 | - | 1 - 2 | 216 | 67 | ||
FQ229157.1 | - | 1 - 2 | 215 | 67 | ||
FQ229247.1 | - | 1 - 2 | 216 | 67 | ||
FQ229569.1 | - | 1 - 2 | 217 | 67 | ||
FQ229616.1 | - | 1 - 2 | 216 | 67 | ||
FQ229633.1 | - | 1 - 2 | 216 | 67 | ||
FQ229689.1 | - | 1 - 2 | 216 | 67 | ||
FQ229758.1 | - | 1 - 2 | 216 | 67 | ||
FQ229908.1 | - | 1 - 2 | 217 | 67 | ||
FQ229911.1 | - | 1 - 2 | 216 | 67 | ||
FQ229916.1 | - | 1 - 2 | 214 | 67 | ||
FQ229923.1 | - | 1 - 2 | 215 | 67 | ||
FQ230003.1 | - | 1 - 2 | 214 | 67 | ||
FQ230018.1 | - | 1 - 2 | 224 | 67 | ||
FQ230328.1 | - | 1 - 2 | 217 | 67 | ||
FQ233024.1 | - | 1 - 2 | 216 | 67 | ||
FQ234673.1 | - | 1 - 2 | 218 | 67 | ||
FQ235255.1 | - | 1 - 2 | 216 | 67 | ||
U03390.1 | - | 1 - 2 | 208 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
