Assay Details
| Gene Symbol: Gstp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012577.2 | NP_036709.1 | 2 - 3 | 105 | 90 | |
| GenBank mRNA | BC058439.1 | - | 2 - 3 | 109 | 6889675 | 90 |
| BC058440.1 | - | 2 - 3 | 109 | 6917961 | 90 | |
| FQ217279.1 | - | 2 - 3 | 105 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC058439.1,BC058440.1,NM_012577.2 transcripts.
This assay is in the 5'UTR region of the BC058439.1,BC058440.1,NM_012577.2 transcripts.
Set Membership: |
|

Gstp1
Genomic Map