Assay Details
| Gene Symbol: Oaz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297557.1 | NP_001284486.1 | 2 - 3 | 371 | 60 | |
| NM_001301048.1 | NP_001287977.1 | 2 - 3 | 365 | 60 | ||
| NM_139081.2 | NP_620781.1 | 2 - 3 | 371 | 60 | ||
| GenBank mRNA | D10706.1 | - | 2 - 3 | 343 | 60 | |
| FQ211375.1 | - | 2 - 3 | 371 | 60 | ||
| FQ213504.1 | - | 2 - 3 | 371 | 60 | ||
| FQ213794.1 | - | 2 - 3 | 371 | 60 | ||
| FQ214019.1 | - | 2 - 3 | 366 | 60 | ||
| FQ214051.1 | - | 2 - 3 | 366 | 60 | ||
| FQ214373.1 | - | 2 - 3 | 366 | 60 | ||
| FQ214458.1 | - | 2 - 3 | 366 | 60 | ||
| FQ216817.1 | - | 2 - 3 | 371 | 60 | ||
| FQ218486.1 | - | 2 - 3 | 366 | 60 | ||
| FQ218990.1 | - | 2 - 3 | 366 | 60 | ||
| FQ220491.1 | - | 2 - 3 | 366 | 60 | ||
| FQ220580.1 | - | 2 - 3 | 371 | 60 | ||
| FQ225116.1 | - | 2 - 3 | 366 | 60 | ||
| FQ225909.1 | - | 2 - 3 | 366 | 60 | ||
| FQ225949.1 | - | 2 - 3 | 369 | 60 | ||
| FQ229954.1 | - | 2 - 3 | 366 | 60 | ||
| FQ231088.1 | - | 2 - 3 | 366 | 60 | ||
| FQ232667.1 | - | 2 - 3 | 366 | 60 | ||
| FQ232997.1 | - | 2 - 3 | 366 | 60 | ||
| FQ233723.1 | - | 2 - 3 | 366 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Oaz1
Genomic Map