Assay Details
| Gene Symbol: Nampt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177928.3 | NP_808789.1 | 5 - 6 | 633 | 63 | |
| GenBank mRNA | AB081730.1 | - | 5 - 6 | 608 | 63 | |
| AY831728.1 | - | 5 - 6 | 606 | 63 | ||
| BC085681.1 | - | 5 - 6 | 633 | 7192109 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nampt
Genomic Map