Assay Details
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181627.2 | NP_853658.1 | 380 | 137 | ||
GenBank mRNA | AY291062.1 | - | 380 | 137 | ||
AY291063.2 | - | 575 | 137 | |||
AY919343.1 | - | 359 | 137 | |||
FQ213378.1 | - | 397 | 137 | |||
FQ213584.1 | - | 397 | 137 | |||
FQ214037.1 | - | 398 | 137 | |||
FQ214498.1 | - | 408 | 137 | |||
FQ229190.1 | - | 397 | 137 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
