Assay Details
| Gene Symbol: Ffar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153304.2 | NP_695216.1 | 1 | 849 | 65 | |
| GenBank mRNA | AB095744.1 | - | 1 | 847 | 65 | |
| AF539810.1 | - | 1 | 851 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ffar1
Genomic Map