Assay Details
| Gene Symbol: Pax9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039539.1 | NP_001034628.1 | 1 - 2 | 173 | 67 | |
| GenBank mRNA | AB248957.1 | - | 1 - 2 | 173 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB248957.1, NM_001039539.1 transcripts.
Set Membership: |
|

Pax9
Genomic Map