Assay Details
| Gene Symbol: Chtop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749811.3 | XP_003749859.2 | 1 | 790 | 56 | |
| Gene Symbol: Chtop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232710.2 | XP_006232772.1 | 5 - 6 | 1645 | 56 | |
| XM_006232711.2 | XP_006232773.1 | 5 - 6 | 1561 | 56 | ||
| XM_006232713.2 | XP_006232775.1 | 5 - 6 | 1557 | 56 | ||
| GenBank mRNA | BC100084.1 | - | 5 - 6 | 593 | 7385093 | 56 |
| FQ231784.1 | - | 4 - 5 | 495 | 56 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chtop
Genomic Map