Assay Details
Gene Symbol: Rsbn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135872.1 | NP_001129344.1 | 5 - 6 | 2034 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
