Assay Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749718.3 | XP_003749766.1 | 6 - 7 | 686 | 59 | |
| XM_008762849.1 | XP_008761071.1 | 6 - 7 | 686 | 59 | ||
More Information
Set Membership: |
|

Slc37a3
Genomic Map