Assay Details
| Gene Symbol: Fam96a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008327.1 | NP_001008328.1 | 1 - 2 | 257 | 69 | |
| GenBank mRNA | BC086524.1 | - | 1 - 2 | 257 | 7303322 | 69 |
| FQ210328.1 | - | 1 - 2 | 238 | 69 | ||
| FQ215407.1 | - | 1 - 2 | 294 | 69 | ||
| FQ216067.1 | - | 1 - 2 | 301 | 69 | ||
| FQ216458.1 | - | 1 - 2 | 295 | 69 | ||
| FQ216583.1 | - | 1 - 2 | 255 | 69 | ||
| FQ218392.1 | - | 1 - 2 | 238 | 69 | ||
| FQ218618.1 | - | 1 - 2 | 301 | 69 | ||
| FQ220244.1 | - | 1 - 2 | 217 | 69 | ||
| FQ225271.1 | - | 1 - 2 | 154 | 69 | ||
| FQ232448.1 | - | 1 - 2 | 255 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam96a
Genomic Map