Assay Details
Gene Symbol: Paip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014148.1 | NP_001014170.1 | 4 - 5 | 460 | 121 | |
XM_008772044.1 | XP_008770266.1 | 2 - 3 | 307 | 121 | ||
XM_008772046.1 | XP_008770268.1 | 2 - 3 | 341 | 121 | ||
XM_008772047.1 | XP_008770269.1 | 3 - 4 | 363 | 121 | ||
XM_008772048.1 | XP_008770270.1 | 3 - 4 | 380 | 121 | ||
XM_008772049.1 | XP_008770271.1 | 2 - 3 | 453 | 121 | ||
GenBank mRNA | BC079261.1 | - | 4 - 5 | 460 | 7133501 | 121 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
