Assay Details
| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053588.2 | NP_446040.1 | 610 | 101 | ||
| GenBank mRNA | AF315468.1 | - | 571 | 101 | ||
| BC061821.1 | - | 610 | 5599087 | 101 | ||
More Information
Set Membership: |
|

Rnf138
Genomic Map