Assay Details
Gene Symbol: LOC100360606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728842.4 | XP_002728888.3 | 6 - 7 | 1838 | 124 | |
XM_006230445.2 | XP_006230507.1 | 7 - 8 | 2003 | 124 | ||
XM_008759959.1 | XP_008758181.1 | 6 - 7 | 2090 | 124 | ||
XM_008759960.1 | XP_008758182.1 | 7 - 8 | 1735 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
